Learn More
Tritech Research Inc NARROW VIAL TRAY/DIVIDER EACH
SDP
Supplier:  Tritech Research Inc T260
Encompass
Narrow vial tray and divider. Cardboard trays with cell dividers. 10 x 10 row configuration for narrow (25mm diameter) or shorty shell vials. 50 per case.
Catalog No. NC9650380
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.