Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Nasal Swab Training Model
SCI_ED

Catalog No. S28932 Shop All Eisco Products
Change view
Click to view available options
Dimensions (L x W x H):
12 x 10 x 8 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S28932 12 x 10 x 8 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S28932 Supplier Eisco™ Supplier No. BI201001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

12 in. nasal swab training model for the demonstration of nasopharyngeal testing.

  • Visual demonstration of a nasopharyngeal swab test used for testing for many respiratory viruses and airborne diseases
  • Fully three-dimensional with internal anatomy shown on cross-section
  • Made of polyvinyl chloride (PVC) plastic for durability and mounted onto a stand
  • Suitable for many environments including educational and medical uses
  • A great addition to a laboratory, a doctor's office, or a science classroom
  • Designed by subject matter experts and hand-painted using vibrant, eco-friendly paint
  • Model with stand measures 12 x 10 x 8 in.

Specifications

Dimensions (L x W x H) 12 x 10 x 8 in.
For Use With (Application) Visual Demonstration of a Nasopharyngeal Swab Test
Material PVC
Mounting Stand
Product Type Model, Training
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.