Learn More
Nasco CPR Prompt™ Training and Practice Manikin - TPAK 100 Adult/Child
SDPSCI_ED
Catalog No. S97921 Shop All Simulaids, Inc. Products
Click to view available options
Economical package
Catalog No.
S97921 Economical package
1 options
Catalog No. S97921 Supplier Nasco Supplier No. LF06700
Includes
Includes: Each set is packed in a rugged nylon carrying case with a supply of 50 face-shield lung bags and an insertion tool
Design for practice of Heimlich maneuver and abdominal thrusts, as well as CPR and mouth-to-mouth breathing.
Realistic anatomical markings (Adam's apple, carotid arteries, navel, rib cage, chest notch) make it easy to learn correct hand placement for checking pulse, performing chest compressions, and more.
  • Head tilt/chin lift required to open airway, lung bags are replaceable
  • Proper anatomical landmarks
  • Audible compression clicker to ensure proper technique
  • Easy-to-clean heads are dishwasher safe and can easily be sanitized
  • Designed to meet American Heart Association criteria for desirable manikin features
  • With age selector dial that adjusts chest compression piston for learning adult or child techniques
Ideal for group training – allows each student to have their own manikin for maximum practice time
Warranty and Services
Warranty: 3 year warranty
Specifications
Product Type Training and Practice Manikin
For Use With (Application) Group Training Allows each Student to have their own Manikin for Maximum Practice Time
Includes Five adult/child manikins, two infant manikins, 50 adult/child face shield/lung bags, 20 infant face shield/lung bags and nylon carry case