Learn More
Whirl-Pak™ Sterilized Spoons
GREENER_CHOICESDP
Catalog No. 0181282 Shop All Whirl-Pak Products
Change view
Click to view available options
Material:
Polystyrene
Catalog No. Material
01-812-82 Polystyrene
1 options
Catalog No. 01-812-82 Supplier Whirl-Pak™ Supplier No. B01041A
Add to Cart
Add to Cart
Gas-sterilized spoons are ideal for use with Whirl-Pak bags where sterile techniques are required for collection of samples.
Gas-sterilized spoons are ideal for use with Nasco Whirl-Pak™ bags where sterile techniques are required for collection of samples
  • Easy-to-use spoons
  • Polystyrene
  • Sterile
Specifications
Disposable Single-use
Product Type Spoon
For Use With (Equipment) Whirl-Pak™ bags
For Use With (Application) Nasco Whirl-pak bags where sterile techniques are required for collection of samples
Material Polystyrene
Sterility Sterile
Autoclavable Not Autoclavable
Shape Spoon