Learn More
3M™ Nassau™ Plus™ Eyewear
Catalog No. 17028F Shop All 3M Products
Change view
Click to view available options
Frame Color:
Red, White, Blue
Catalog No. Frame Color
17-028F Red, White, Blue
1 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations and the user instructions, warnings, and limitatins accompanying each product.
Catalog No. 17-028F Supplier 3M™ Supplier No. 7010385305
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations and the user instructions, warnings, and limitatins accompanying each product.
With sporty styling and rugged construction, these spectacles provide a high fashion look in eye protection
  • Lightweight but durable
  • Frame features adjustable temples for a custom fit
  • Integrated sideshield for wraparound protection
  • Single-piece high-impact polycarbonate lens
  • Comfortable flared nosepiece with high set bridge option ideal for broad or low nasal profile
  • DX™ antifog hard coat protects lens from fog, scratching, and most chemicals
  • 3M Catalog# 14300-00000-20 , 3M Catalog# 14327-00000-20
Compliance
Meets ANSI Z87.1-2003 and CSA Z94.3-2007 standards
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Clear
Frame Color Red, White, Blue
Model 14327-00000-20
Lens Material Polycarbonate