Learn More
Yamato Scientific America, Inc. Yamato 99L Natural Convection Oven, Programmable, 115V
Supplier:  Yamato Scientific America, Inc. DVS402C
Encompass_Preferred
Programmable 99L natural convection constant temperature drying oven incorporating programmable operation functions and enhanced safety features and operability. Excellent temperature accuracy. Easy to use and maintain. Equipped with a 6 pattern PID program controller with easy program settings (30 steps x 1, 15 steps x 2, 10 steps x 3). Simultaneous display of set constant and measured temperature. Operating temp ambient +5~260°C
Catalog No. 50-189-7981
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.