Learn More
Pickering Laboratories Inc NAT SAMP AA STD ACED 0.1N HCL
SDP
Supplier:  Pickering Laboratories Inc 17000180
Encompass_Preferred
Native Sample Standard, Acidics and Neutrals, in 0.1 N HCl, 2.5 µmole/mL, Cystine 1.25 µmole/mL, each (5 mL /bottle) Amino Acid Analysis.
Catalog No. NC9054999
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.