Learn More
Invitrogen™ NAT11 Polyclonal Antibody

Description
A suggested positive control is human thymus tissue lysate. PA5-20882 can be used with blocking peptide PEP-0996.
Specifications
Specifications
| Antigen | NAT11 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | NAA40 |
| Gene Accession No. | Q86UY6, Q8VE10 |
| Gene Alias | 4931433E08Rik; AU023197; N(alpha)-acetyltransferase 40, NatD catalytic subunit; N(alpha)-acetyltransferase 40, NatD catalytic subunit, homolog; N(alpha)-acetyltransferase 40, NatD catalytic subunit, homolog (S. cerevisiae); Naa40; N-acetyltransferase 11; N-acetyltransferase 11 (GCN5-related, putative); N-alpha acetyl transferase 40; Nalpha acetyltransferase 40; N-alpha-acetyltransferase 40; N-alpha-acetyltransferase 40, NatD catalytic subunit; N-alpha-acetyltransferase D; NAT11; NatD; natD catalytic subunit; PATT1; Protein acetyltransferase 1 |
| Gene Symbols | NAA40 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.