Learn More
NAT8 Mouse anti-Human, Clone: OTI5A8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500544

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This protein, isolated using the differential display method to detect tissue-specific genes, is specifically expressed in kidney and liver. The encoded protein shows amino acid sequence similarity to N-acetyltransferases. A similar protein in Xenopus affects cell adhesion and gastrulation movements, and may be localized in the secretory pathway. A highly similar paralog is found in a cluster with this gene.Specifications
NAT8 | |
Monoclonal | |
Unconjugated | |
NAT8 | |
ATase2, CCNAT, CML1, GLA, Hcml1, TSC501, TSC510 | |
Mouse | |
Affinity Chromatography | |
RUO | |
9027 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI5A8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9UHE5 | |
NAT8 | |
Full-length protein expressed in 293T cell transfected with human NAT8 expression vector | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.