Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAT8B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577166
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577166 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577166 Supplier Invitrogen™ Supplier No. PA577166
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

The protein encoded by this gene is highly similar to the N-acetyltransferase 8 (NAT8) gene product, which is a kidney and liver protein with homology to bacterial acetyltransferases involved in drug resistance. This gene is localized on chromosome 2 in the vicinity of the NAT8 gene and may represent a pseudogene of NAT8. This gene contains two polymorphic nonsense mutations that disrupt the active site of the protein. The full-length product of this gene contains a complete acetyltransferase domain and is identical in length to NAT8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAT8B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene NAT8B
Gene Accession No. E0CYC6, Q9UHF3
Gene Alias Acetyltransferase 1; ATase1; camello-like 1 like; camello-like protein 1; Camello-like protein 2; camello-like protein 6; Cml1; CML2; Cml6; Hcml2; N-acetyltransferase 8B; N-acetyltransferase 8B (GCN5-related, putative, gene/pseudogene); N-acetyltransferase 8B (putative, gene/pseudogene); N-acetyltransferase Camello 2; Nat8; Nat8b; NAT8BP; probable N-acetyltransferase 8B; probable N-acetyltransferase CML6; Putative N-acetyltransferase 8B; RGD621605
Gene Symbols NAT8B, Nat8b-ps
Host Species Rabbit
Immunogen Recombinant full length Human NAT8B.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171084, 434057, 51471
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.