Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAT8B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981499
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1499 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1499 Supplier Bioss Supplier No. BS11592R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is highly similar to the N-acetyltransferase 8 (NAT8) gene product, which is a kidney and liver protein with homology to bacterial acetyltransferases involved in drug resistance. This gene is localized on chromosome 2 in the vicinity of the NAT8 gene and may represent a pseudogene of NAT8. This gene contains two polymorphic nonsense mutations that disrupt the active site of the protein. The full-length product of this gene contains a complete acetyltransferase domain and is identical in length to NAT8.

Specifications

Antigen NAT8B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NAT8B
Gene Accession No. Q9UHF3
Gene Alias Acetyltransferase 1; ATase1; camello-like 1 like; Camello-like protein 1; Camello-like protein 2; Camello-like protein 6; Cml1; CML2; Cml6; Hcml2; N-acetyltransferase 8B; N-acetyltransferase 8B (GCN5-related, putative, gene/pseudogene); N-acetyltransferase 8B (putative, gene/pseudogene); N-acetyltransferase Camello 2; Nat8; NAT8B; NAT8BP; probable N-acetyltransferase 8B; Probable N-acetyltransferase CML6; Putative N-acetyltransferase 8B; RGD621605
Gene Symbols NAT8B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NAT8B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51471
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.