Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits

Catalog No. 0337538C
Change view
Click to view available options
Color:
Amber
Clear
Closure Type:
Polypropylene
Polypropylene, Flanged
Septum:
PTFE/Silicone
Packaging Type:
Polybag
Storage Tray
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Type Septum Packaging Type
03-375-38C Clear Polypropylene PTFE/Silicone Polybag
03-376-460 Amber Polypropylene, Flanged PTFE/Silicone Storage Tray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-375-38C Supplier Thermo Scientific™ Supplier No. C4013015A

Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.

Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.

  • Packaged in convenient vial trays with clear covers or in economical polybags
  • Vial trays are stackable for easy storage
  • All caps are manufactured from virgin polypropylene unless noted

Includes: 

100 vials with pre-attached caps and septa

TRUSTED_SUSTAINABILITY

Specifications

Closure Color Black
Color (Seal) Red/White
Description Clear Glass; PP Black Cap; Red PTFE/White Silicone septum; in polybag
Color Clear
Material Glass, PTFE, Silicone
Volume (Metric) 2 mL
Septum PTFE/Silicone
Packaging Type Polybag
Closure Material Polypropylene
Closure Type Polypropylene
Closure Size 8-425
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.