Learn More
National Diagnostics™ Calci-Clear™ Decalcifying Solution
Description
- Moderate speed decalcifier
- Mildness balanced with performance
- Reduces interference with subsequent staining
- Minimizes tissue distortion and structure loss
- May be used to decalcify any tissue
Specifications
Specifications
| Quantity | 1 gal. |