Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ National Economy Vials

Catalog No. 03250609
Change view
Click to view available options
Color:
Amber
Clear
Product Type:
Clear
Crimp Top Vial
Screw Thread Vial
Volume (Metric) Working:
2 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type Volume (Metric) Working
03-250-609 Clear Crimp Top Vial 2 mL
03-250-610 Amber Crimp Top Vial 2 mL
03-250-611 Clear Clear 2 mL
03-250-623 Amber Screw Thread Vial 2 mL
03-250-638 Clear Screw Thread Vial 2 mL
03-250-639 Amber Screw Thread Vial 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 03-250-609 Supplier Thermo Scientific™ Supplier No. 60180701

Thermo Scientific™ National Economy Vials are offered as clear or amber, in a range of styles to meet specific application needs.

Thermo Scientific™ National Economy Vials are offered as clear or amber, in a range of styles to meet specific application needs.

TRUSTED_SUSTAINABILITY

Specifications

Description 60180-701
Color Clear
Volume (Metric) 2 mL
Volume (Metric) Working 2 mL
For Use With (Equipment) Thermo Scientific, Agilent, PE, Shimadzu, Varian
Product Type Crimp Top Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.