Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Biohazard Floor Signs

Catalog No. 179862E Shop All National Marker Company Products
Change view
Click to view available options
Dimensions (L x W):
20 x 12 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
17-986-2E 20 x 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-986-2E Supplier National Marker™ Supplier No. FS 11

missing translation for 'validMainframeMsg'

Specifications

Material Polyethylene
Quantity 1 Ea.
Height (English) 20 in.
Width (English) 12 in.
Dimensions (L x W) 20 x 12 in.
Displayed Text BIOHAZARD AUTHORIZED PERSONNEL ONLY
Product Type Warning Sign
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.