Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Biohazard Labels

Catalog No. 1798690D Shop All AccuformNMC Products
Change view
Click to view available options
Display Text:
Biohazard Label
Biohazard Label + Hazard ID
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text
17-986-90D Biohazard Label + Hazard ID
17-986-90F Biohazard Label
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-986-90D Supplier National Marker™ Supplier No. WOL5

Small labels serving as visual reminders of biohazards

  • Material: Pressure-sensitive paper
  • Size: 2.54 × 7.62cm (1 × 3 in.)
  • Color: Orange with black imprint

Specifications

Certifications/Compliance OSHA 29 CFR 1910.145
Color Orange
Format Label
Material Paper
Product Type Biohazard: Hazard Identity and write-on space
Display Text Biohazard Label + Hazard ID
For Use With (Application) Biohazard
Quantity 1 Roll
Size (English) 1 x 3 in.
Size (Metric) 2.54 x 7.62 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.