Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Biohazard: Peligro Biologico Sign

Catalog No. 19035049 Shop All AccuformNMC Products
Change view
Click to view available options
Material:
Plastic
Vinyl
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-035-049 Plastic
19-035-050 Vinyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-035-049 Supplier National Marker™ Supplier No. S91R

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

Spanish language biohazard warning sign

  • Orange sign with black text
  • Sign: 17.7 × 17.7cm (7 × 7 in.)
  • Available in plastic or self-sticking polyester

Specifications

Material Plastic
Quantity 1 Ea.
Height (English) 7 in.
Width (English) 7 in.
Displayed Text BIOHAZARD/PELIGRO BIOLOGICO
Product Type Biohazard Sign
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.