Learn More
National Marker™ Danger: Confined Space Entry Permit Not Required Signs
Shop All AccuformNMC ProductsSpecifications
Specifications
| Certifications/Compliance | OSHA 29 CFR 1910.145 |
| Material | Plastic |
| Quantity | 1 Ea. |
| For Use With (Application) | Perfect way to alert employees and guests when a hazard is present in your facility |
| Height (English) | 10 in. |
| Max. Temperature (English) | 183°F |
| Max. Temperature (Metric) | 84°C |
| Thickness | 0.05 in. |
| Width (English) | 14 in. |
| Displayed Text | DANGER Confined Space Entry Permit not Required |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.