Learn More
National Marker™ Danger: Follow Confined Space Entry Procedure Before Signs
Shop All AccuformNMC ProductsSpecifications
Specifications
| Certifications/Compliance | OSHA 29 CFR 1910.145 |
| Material | Plastic |
| Quantity | 1 Ea. |
| Description | Rigid Plastic |
| For Use With (Application) | Perfect way to alert employees and guests when a hazard is present in your facility |
| Height (English) | 7 in. |
| Max. Temperature (English) | 183°F |
| Max. Temperature (Metric) | 84°C |
| Thickness | 0.05 in. |
| Width (English) | 10 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.