Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Danger Laser Sign

Catalog No. 1798540C Shop All National Marker Company Products
Catalog No. 17-985-40C Supplier National Marker™ Supplier No. D 136 R

missing translation for 'validMainframeMsg'

Alert semployees and guests of hazards relating to lasers in the facility

  • Made of rigid plastic
  • Size: 17.7 × 25.4cm (7 × 10 in.)

Compliance

Signs comply with OSHA 29 CFR 1910.145

Specifications

Certifications/Compliance OSHA 29 CFR 1910.145
Material Plastic
Quantity 1 Ea.
Description Rigid Plastic
For Use With (Application) Indoor and outdoor use
Height (English) 7 in.
Max. Temperature (English) 183°F
Max. Temperature (Metric) 84°C
Thickness 0.05 in.
Width (English) 10 in.
Displayed Text DANGER Laser
Product Type OSHA Sign
Color Red on White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.