Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ DOT Shipping Labels

Catalog No. 17987170G Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 Ea.
25/Pk.
500/Pk.
Display Text:
Corrosive
Corrosives 8
Flammable Gas
Flammable Liquid
Flammable Liquid 3
Flammable Solid
Flammable Solids
Non Flammable Gas
Non-flammable Gas 2
Organic Peroxide
Oxidizers And organic peroxides
Radioactive I
Radioactive II
Material:
Paper
Vinyl
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Display Text Material
17-987-170G 500/Pk. Corrosive Paper
17-981-56H 25/Pk. Corrosives 8 Vinyl
17-985-411F 25/Pk. Organic Peroxide Vinyl
17-987-170B 25/Pk. Flammable Liquid Vinyl
17-981-56D 25/Pk. Flammable Liquid 3 Paper
17-987-170C 1 Ea. Non Flammable Gas Paper
17-987-170F 500/Pk. Flammable Solid Paper
17-981-56G 25/Pk. Flammable Solids Vinyl
17-981-56J 25/Pk. Oxidizers And organic peroxides Vinyl
17-987-170L 500/Pk. Radioactive I Paper
17-987-170A 500/Pk. Flammable Gas Paper
17-987-170M 500/Pk. Radioactive II Paper
17-981-56F 25/Pk. Non-flammable Gas 2 Vinyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 17-987-170G

For marking packages with hazardous materials

  • Help to meet the requirements in 49CFR for P.O.P. (“Performance Oriented Packaging”) labeling of hazardous material shipments
  • Must be retained on packaging until it is sufficiently cleaned of residue and purged of vapors

Order Info

Measure 4 x 4 in. . Availalble in pressure-sensitive paper or vinyl.

Specifications

Certifications/Compliance DOT 49 CFR 172.519
Format Label
Material Paper
Product Type DOT Shipping Label
Display Text Corrosive
For Use With (Application) Designed specifically for vehicles dedicated to the repeat transportation of a single material
Quantity 500/Pk.
Width (English) 4 in.
Height (English) 4 in.
Height (Metric) 10.16 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.