Learn More
Description
- Help to meet the requirements in 49CFR for P.O.P. (“Performance Oriented Packaging”) labeling of hazardous material shipments
- Must be retained on packaging until it is sufficiently cleaned of residue and purged of vapors
Order Info
Measure 4 x 4 in. . Availalble in pressure-sensitive paper or vinyl.
Specifications
Specifications
| Certifications/Compliance | DOT 49 CFR 172.407 |
| Format | Label |
| Material | Paper |
| Product Type | DOT Shipping Label |
| Display Text | Radioactive I |
| For Use With (Application) | Hazard |
| Quantity | 500/Pk. |
| Width (English) | 4 in. |
| Height (English) | 4 in. |
| Height (Metric) | 10.16 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.