Learn More
National Marker™ Hazardous Material Pressure Sensitive Numbers for Labeling
Shop All AccuformNMC ProductsSpecifications
Specifications
| Color | Black, White |
| Format | Label |
| Material | Vinyl |
| Product Type | Label Number |
| Display Text | 1 |
| For Use With (Application) | Hazardous materials |
| Quantity | 5/Pk. |
| Thickness (English) | 0.0045 mil |
| Width (English) | 5 in. |
| Height (English) | 5 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.