Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Hazardous Materials Classification Reference Signs

Catalog No. 179844B Shop All AccuformNMC Products
Change view
Click to view available options
Material:
Plastic
Vinyl
Width (English):
2.5 in.
8 in.
10 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Width (English)
17-984-4B Plastic 8 in.
17-984-4C Plastic 10 in.
17-984-4A Plastic 2.5 in.
17-985-34A Vinyl 8 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-984-4B Supplier National Marker™ Supplier No. HMC8R

Ideal for training or quick reference

  • Easy-to-understand reference signs
  • Graphically explain the NFPA hazard identification coding system.
  • Available in 0.03 in. thick rigid plastic with predrilled mounting holes, or in 4mil thick pressure-sensitive vinyl

Specifications

Material Plastic
Quantity 1 Ea.
Description Rigid Plastic
For Use With (Application) Training or quick reference, graphically explain the NFPA hazard identification coding system
Height (English) 11 in.
Thickness 0.03 in.
Width (English) 8 in.
Displayed Text Hazardous Materials Classification Reference
Product Type Hazardous Materials Classification Reference Sign
Color Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.