Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Paper Shipping Labels for Hazardous and Nonhazardous Waste

Catalog No. 1798162C Shop All AccuformNMC Products
Change view
Click to view available options
Display Text:
Hazardous Waste
Hazardous Waste (California)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text
17-981-62C Hazardous Waste
17-985-409D Hazardous Waste (California)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-981-62C Supplier National Marker™ Supplier No. HW10

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

Chemical- and weather-resistant labels

Order Info

Measure 6 x 6 in.; Pressure-sensitive paper; Roll of 500

Specifications

Certifications/Compliance EPA and DOT, 17-985-409C or -409D, 17-985-409E
Color Yellow
Format Label
Material Paper
Product Type Container Label
Display Text Hazardous Waste
For Use With (Application) Hazardous waste
Quantity 500/Pk.
Thickness (English) 4 mil
Width (English) 6 in.
Height (English) 6 in.
Height (Metric) 15.24 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.