Learn More
Description
Provides a central location for OSHA information on health, flammability, reactivity, and target organ effects. High-impact plastic station comes with attached polypropylene binder for Material Safety Data Sheets, one employee instruction manual and pockets for pocket reference guides. Many companies display their own company-specific guides.
Order Info
To purchase the guides shown in photo or additional instructional manuals, contact your Fisher Safety Customer Service Representative for details.
Specifications
Specifications
| Format | Binder |
| Product Type | MSDS Binder |
| For Use With (Application) | Provides workers with easy access to material safety data sheets with the right-to-know compliance center |
| Subject | Safety and Training Materials |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.