Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Right-To-Know Labels

Catalog No. 1798536C Shop All AccuformNMC Products
Change view
Click to view available options
Display Text:
0
2
3
ACID
ALK
COR
Use No Water graphic (W with slash)
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text
17-985-36C 2
17-985-35A Use No Water graphic (W with slash)
17-985-35C ACID
17-985-35D ALK
17-985-35E COR
17-985-36A 0
17-985-36D 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17-985-36C Supplier National Marker™ Supplier No. DCN62

Diamond-shaped RTK decals with a variety of messages

  • NFPA diamond labels
  • Pressure-sensitive vinyl material
  • Black text on white background

Specifications

Color Black, White
Format Label
Material Vinyl
Product Type Label Number
Display Text 2
For Use With (Application) Right-To-Know
Quantity 5/Pk.
Thickness (English) 0.0045 mil
Width (English) 6 in.
Height (English) 6 in.
Height (Metric) 15.24 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.