Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Vinyl Shipping Labels for Hazardous and Nonhazardous Waste

Catalog No. 1798162B Shop All AccuformNMC Products
Change view
Click to view available options
Color:
Blue
Green
Yellow
Display Text:
Caution Contains PCBs
Hazardous Waste
Hazardous Waste (California)
Hazardous Waste (For Solids)
Hazardous Waste (New Jersey)
Hazardous waste (for liquids)
Non-Hazardous Waste
Non-Regulated Waste
Thickness (English):
0.002 mil
0.0045 mil
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Display Text Thickness (English)
17-981-62B Yellow Caution Contains PCBs 0.0045 mil
17-981-62A Yellow Hazardous Waste 0.002 mil
17-985-409C Yellow Hazardous Waste (California) 0.002 mil
17-985-409E Yellow Hazardous Waste (New Jersey) 0.0045 mil
17-985-409A Green Non-Hazardous Waste 0.0045 mil
17-985-409J Yellow Hazardous waste (for liquids) 0.0045 mil
17-985-409B Blue Non-Regulated Waste 0.0045 mil
17-985-409H Yellow Hazardous Waste (For Solids) 0.0045 mil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-981-62B Supplier National Marker™ Supplier No. HW4C

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical- and weather-resistant labels

Order Info

Measure 6 x 6 in.; 4mil pressure-sensitive vinyl; In packs of 25

Specifications

Certifications/Compliance EPA and DOT, 17-985-409C or -409D, 17-985-409E
Color Yellow
Format Label
Material Vinyl
Product Type Shipping Label
Display Text Caution Contains PCBs
For Use With (Application) Suitable for both indoor and outdoor applications
Quantity 25/Pk.
Thickness (English) 0.0045 mil
Width (English) 6 in.
Height (English) 6 in.
Height (Metric) 15.24 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.