Learn More
National Marker™ Vinyl Shipping Labels for Hazardous and Nonhazardous Waste
Shop All AccuformNMC ProductsDescription
Order Info
Measure 6 x 6 in.; 4mil pressure-sensitive vinyl; In packs of 25
Specifications
Specifications
| Certifications/Compliance | EPA and DOT, 17-985-409C or -409D, 17-985-409E |
| Color | Green |
| Format | Label |
| Material | Vinyl |
| Product Type | Shipping Label |
| Display Text | Non-Hazardous Waste |
| For Use With (Application) | Suitable for both indoor and outdoor applications |
| Quantity | 25/Pk. |
| Thickness (English) | 0.0045 mil |
| Width (English) | 6 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.