Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Write-On Hazard Warning Labels

Catalog No. 1798690C Shop All AccuformNMC Products
Change view
Click to view available options
Display Text:
Biohazard Infectious Waste
Hazard Warning
NFPA
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text
17-986-90C NFPA
17-986-90E Biohazard Infectious Waste
17-986-90B Hazard Warning
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-986-90C Supplier National Marker™ Supplier No. WOL4

National Marker Write-On Hazard Warning Labels

Specifications

Format Label
Material Paper
Product Type Label
Display Text NFPA
For Use With (Application) Hazard warning, NFPA
Quantity 500/Pk.
Width (English) 3 in.
Height (English) 1 in.
Height (Metric) 2.54 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.