Learn More
Description
- Pressure-sensitive vinyl labels
- Write on and/or use the Personal Protection Numbers and Symbols (Cat. No. 17-981-61D)
- Measure 6 × 6 in. (15.2 × 15.2cm)
Order Info
Available in pack of 10.
Compliance
Comply with 29 CFR1910.1200.
Specifications
Specifications
| Certifications/Compliance | 29 CFR 1910.1200 |
| Color | Red, Yellow, Blue, White |
| Format | Label |
| Material | Vinyl |
| Product Type | Label |
| For Use With (Application) | Hazardous materials |
| Quantity | 5/Pk. |
| Width (English) | 15 in. |
| Height (English) | 15 in. |
| Height (Metric) | 38.1 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.