Learn More
Description
- Identify specific hazards, as well as the degree of health hazard, flammability, and reactivity associated with chemicals
- Simply determine the hazards of a specific chemical and write the appropriate NFPA hazard number in the corresponding colored area
Order Info
Two sizes: 2 x 2 in. (5 x 5cm) and 4 x 4 in. (10 x 10cm). Available in rolls of 250 pressure-sensitive paper labels per roll.
Specifications
Specifications
| Color | Red, Yellow, Blue, White |
| Format | Label |
| Material | Paper |
| Product Type | Label |
| Display Text | NFPA |
| For Use With (Application) | Use in laboratories and other indoor work areas |
| Quantity | 250/Pk. |
| Width (English) | 2 in. |
| Height (English) | 2 in. |
| Height (Metric) | 5.08 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.