Learn More
Thermo Scientific™ National Polypropylene and Glastic™ Vials with 8-425 Finish
Catalog No. 03340261
Change view
Click to view available options
Volume (Metric) Working:
400 μL
Catalog No. Volume (Metric) Working
03-340-261 400 μL
1 options
Catalog No. 03-340-261 Supplier Thermo Scientific™ Supplier No. C401319
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Thermo Scientific™ National Polypropylene and Glastic Vials provide an economical alternative to glass for some applications.
Thermo Scientific™ National Polypropylene and Glastic Vials provide an economical alternative to glass for some applications.
  • Precision formed tips are designed to aid sample recovery for limited-volume applications
  • Glastic vial features a glass insert pre-inserted inside a clear TPX vial
TRUSTED_SUSTAINABILITY
Specifications
Description 600µL MicroVials
Color Clear
Material Polypropylene
Volume (Metric) 600 μL
Volume (Metric) Residual <4 μL
Volume (Metric) Working 400 μL
Closure Size 8-425