Learn More
National Scientific™ BioStor™ Screw Cap Vials, Skirted
Description
Multi-purpose design serves a broad spectrum of bio-technology laboratory requirements including the collection, processing, centrifugation, long-term storage, and OEM packaging/shipping of restriction enzymes or other DNA/RNA modifying reagents.
- Circular 3mm syringe port in cap which is easily pierced for radioisotope labeling and fraction collecting
- Inert, LN2-grade polypropylene protects critical sample viability at extreme temperatures ranging from +121C to -195C (LN2 vapor phase)
- Two-turn closure design ensures that tightening of unique screw-cap will uniformly compress “O”-ring against beveled seat, eliminating distortion and maintaining seal integrity
- Vials, caps and ethylene/propylene “O”-rings chemial-resistant and autoclavable with cap loosened
- Molded-in volume lines and etched writing panels on vials and caps will not crack off or dissolve
- External ribs on vials position and lock into serrated holes in metal BioStor Racks, allowing single-handed opening and closing of screw-caps
- Available in natural and ainbow pack
Specifications
Specifications
| Color | Natural |
| Sterility | Sterile |
| Volume (Metric) | 1.6 mL |
| For Use With (Application) | Collection, processing, centrifugation, long term storage and OEM packaging or shipping of restriction enzymes or other DNA or RNA modifying reagents |
| Includes | Reinforced support collar, flexible ethylene or propylene O-ring, external rib, circular 3mm syringe port |
| Temperature Range (English) | -319°F to +249°F |
| Temperature Range (Metric) | -195°C to +121°C |
| Volume (English) | 0.05 oz. |
| Closure Type | Screw Cap |
| Quantity | 500/Cs. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.