Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Scientific™ BioStor™ Screw Cap Vials, Skirted

Catalog No. 1184415
Change view
Click to view available options
Volume (Metric):
1.6 mL
2 mL
Color:
Natural
Rainbow
Sterility:
Non-sterile
Sterile
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color Sterility
11-844-15 1.6 mL Rainbow Sterile
11-844-14 1.6 mL Natural Sterile
11-844-18 2 mL Natural Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-844-15 Supplier National Scientific™ Supplier No. BS16AS PS

Thick-walled construction, with reinforced support collar, allows high speed centrifugation to 17,000xg

Multi-purpose design serves a broad spectrum of bio-technology laboratory requirements including the collection, processing, centrifugation, long-term storage, and OEM packaging/shipping of restriction enzymes or other DNA/RNA modifying reagents.

  • Circular 3mm syringe port in cap which is easily pierced for radioisotope labeling and fraction collecting
  • Inert, LN2-grade polypropylene protects critical sample viability at extreme temperatures ranging from +121C to -195C (LN2 vapor phase)
  • Two-turn closure design ensures that tightening of unique screw-cap will uniformly compress “O”-ring against beveled seat, eliminating distortion and maintaining seal integrity
  • Vials, caps and ethylene/propylene “O”-rings chemial-resistant and autoclavable with cap loosened
  • Molded-in volume lines and etched writing panels on vials and caps will not crack off or dissolve
  • External ribs on vials position and lock into serrated holes in metal BioStor Racks, allowing single-handed opening and closing of screw-caps
  • Available in natural and ainbow pack

Specifications

Color Rainbow
Sterility Sterile
Volume (Metric) 1.6 mL
For Use With (Application) Collection, processing, centrifugation, long term storage and OEM packaging or shipping of restriction enzymes or other DNA or RNA modifying reagents
Includes Reinforced support collar
Temperature Range (English) -319°F to +249°F
Temperature Range (Metric) -195°C to +121°C
Volume (English) 0.05 oz.
Closure Type Screw Cap
Quantity 500/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.