Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Scientific™ MicroTube™ Rack System

Catalog No. 1184285
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Quantity:
10/Pk.
120/Pk.
80/Pk.
960/Pk.
Product Type:
12-strip Microtubes, 1.2mL 12-strips
8-strip Microtubes, 1.2mL 8-strips
Microtubes
Racked MicroTubes, 1.2mL 12-strips
Racked MicroTubes, 1.2mL 8-strips
Racked MicroTubes, 1.2mL Singles
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Quantity Product Type
11-842-85 Non-sterile 120/Pk. 8-strip Microtubes, 1.2mL 8-strips
11-842-84 Non-sterile 960/Pk. Microtubes
11-842-90 Non-sterile 960/Pk. Racked MicroTubes, 1.2mL Singles
11-842-93 Sterile 960/Pk. Racked MicroTubes, 1.2mL 8-strips
11-842-94 Non-sterile 10/Pk. Racked MicroTubes, 1.2mL 12-strips
11-842-86 Non-sterile 80/Pk. 12-strip Microtubes, 1.2mL 12-strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 11-842-85 Supplier National Scientific™ Supplier No. TN0946 08B

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

Featuring a rack with an 8 x 12 hole-pattern on traditional 9mm centers, the entire System conforms to the SBS footprint/configuration compatible with robotic workstations, multichannel pipettors and auto-sampling instruments. An alpha-numeric, non-reversing rack and lid ensure positive sample identification. The lid, with individual condensation rings above each tube, prevents cross-contamination. The inert polypropylene tubes, rack and lid can be autoclaved, centrifuged at moderate speeds and stacked for long-term, low temperature storage of biological samples. The System is ideal for:

  • Bacterial and hybridoma cell uptake
  • Cell harvesting
  • CMV, HSV, HTLV-lll testing
  • Receptor binding assays
  • RIA/EIA serial dilution procedures
  • Recombinant DNA work
  • Pharmaceutical and human genome studies
  • Sample storage librariesRacks, with SBS footprint, feature 8 x 12 hole pattern on 9mm center. making compatible with robotic workstations & multichannel pipettors.

Specifications

Product Type 8-strip Microtubes, 1.2mL 8-strips
Description MicroTube Dimensions: 9mm dia., 46mm height
Material Polypropylene
Sterility Non-sterile
Quantity 120/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.