Learn More
National Scientific™ MicroTube™ Rack System
Description
Featuring a rack with an 8 x 12 hole-pattern on traditional 9mm centers, the entire System conforms to the SBS footprint/configuration compatible with robotic workstations, multichannel pipettors and auto-sampling instruments. An alpha-numeric, non-reversing rack and lid ensure positive sample identification. The lid, with individual condensation rings above each tube, prevents cross-contamination. The inert polypropylene tubes, rack and lid can be autoclaved, centrifuged at moderate speeds and stacked for long-term, low temperature storage of biological samples. The System is ideal for:
- Bacterial and hybridoma cell uptake
- Cell harvesting
- CMV, HSV, HTLV-lll testing
- Receptor binding assays
- RIA/EIA serial dilution procedures
- Recombinant DNA work
- Pharmaceutical and human genome studies
- Sample storage librariesRacks, with SBS footprint, feature 8 x 12 hole pattern on 9mm center. making compatible with robotic workstations & multichannel pipettors.
Specifications
Specifications
| Product Type | Racked MicroTubes, 1.2mL 8-strips |
| Description | MicroTube Dimensions: 9mm dia., 46mm height |
| Holds | Tubes (1.2 mL) |
| Material | Polypropylene |
| Sterility | Sterile |
| Quantity | 960/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.