Learn More
Thermo Scientific™ National Target™ Snap Cap Wide-Opening Vials
Catalog No. 03376459
Change view
Click to view available options
Product Type:
Micro-V™ Amber Glass Snap-It Microsampling Vial
Micro-V™ Clear Glass Snap-It Microsampling Vial
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
03-376-459 Micro-V™ Amber Glass Snap-It Microsampling Vial
03-396-75 Micro-V™ Clear Glass Snap-It Microsampling Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-376-459 Supplier Thermo Scientific™ Supplier No. C4011V6
Thermo Scientific™ National Target Snap Cap Wide-Opening Vials are easier to fill and provide a larger target area for the autosampler needle.
Thermo Scientific™ National Target Snap Cap Wide-Opening Vials are easier to fill and provide a larger target area for the autosampler needle.
  • 2mL, 12 x 32mm Crimp/Snap Ring Vials
  • Fit rotating or robotic arm autosamplers
  • I-D™ vials feature write-on patch with graduations at 0.5, 1.0 and 1.5mL
  • Target Snap-It™ vials can be used with snap caps or aluminum crimp seal closures
  • Fit most autosamplers that accommodate the 12 × 32mm vial format
  • Superior quality 33 expansion borosilicate clear glass (Type 1, Class A) or 51A amber glass
  • Uniformly flat bottom for security with inserts
TRUSTED_SUSTAINABILITY
Specifications
Description Micro-V Amber Glass Snap-It Microsampling Vial
Volume (Metric) 15 μL
Volume (Metric) Residual 15 μL
Product Type Micro-V™ Amber Glass Snap-It Microsampling Vial