Learn More
Abcam Native Bordetella Pertussis Filamentous Hemagglutinin Protein, Unconjugated, 100 μg
Description
- Protein Name: Native Bordetella pertussis Filamentous Hemagglutinin
- Source: Bordetella pertussis Tohama I
- Form: Liquid
- Formulation: pH 8, containing 50% Glycerol, 47.1% Tris HCl, and 2.9% Sodium chloride
- Tested Applications: ELISA, SDS-PAGE
- Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Protein-based Substrate |
| Target Molecule | Bordetella pertussis Filamentous Hemagglutinin |
| Form | Liquid |
| Shipping Condition | Dry Ice |
| Buffer | 50% glycerol, 47.1% Tris-HCl, 2.9% sodium chloride, pH8 |
| Content And Storage | Store at -80°C. |
| Label or Dye | Unconjugated |
| Formulation | 50% Glycerol, 47.1% Tris HCl, 2.9% Sodium chloride, pH 8 |
| Quantity | 100 μg |
| For Use With (Application) | SDS-PAGE, ELISA |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.