Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Native Bordetella Pertussis Toxin, Unconjugated, Liquid, 100 μg

Catalog No. 502682850
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-268-2850 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-268-2850 Supplier Abcam Supplier No. AB274655100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Native Bordetella pertussis toxin protein derived from Tohama I strain, suitable for ELISA and SDS-PAGE.

  • Tested Applications: ELISA, SDS-PAGE
  • Form: Liquid
  • Storage Buffer: pH 8, 50% Glycerol, 47.1% Tris HCl, 2.9% Sodium chloride
  • Source: Bordetella pertussis Tohama I
  • Conjugation: Unconjugated

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Protein-based Substrate
Shipping Condition Dry Ice
Target Molecule Bordetella Pertussis Toxin (ptxD)
Form Liquid
Label or Dye Unconjugated
Gel Compatibility SDS-PAGE Gel
Buffer 50% glycerol, 47.1% Tris-HCl, 2.9% sodium chloride, pH8
Content And Storage Store at -80°C.
Detection Method Colorimetric
Formulation 50% Glycerol, 47.1% Tris HCl, 2.9% Sodium chloride, pH 8
Quantity 100 μg
For Use With (Application) ELISA, SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.