Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Native Chicken Deglycosylated Avidin, FITC Conjugated, 1 mg

Catalog No. 502707012
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7012 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7012 Supplier Abcam Supplier No. AB1362091MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

FITC-conjugated deglycosylated avidin protein with >95% purity for ELISA, WB, and flow cytometry.

  • Target Protein: Deglycosylated Avidin
  • Expression System: Native
  • Purity: >95%
  • Tested Applications: ELISA, Flow Cyt, WB
  • Form: Liquid
  • Buffer: PBS, pH 7.4 with 0.1% Sodium azide

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Blue Ice
Target Molecule Biotin
Reactivity Biotin
Label or Dye FITC
Concentration 1 mg/mL
Buffer PBS, 0.1% sodium azide, pH7.4
Content And Storage Store at 4°C.
Form Liquid
Detection Method Fluorescence
Formulation PBS, 0.1% Sodium azide, pH 7.4
Quantity 1 mg
For Use With (Application) ELISA, Flow Cytometry, Western Blot
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.