Learn More
Abcam Native Chicken Deglycosylated Avidin Protein, Alkaline Phosphatase, 1 mL
Description
- Species: Chicken
- Expression System: Native
- Purity: >90%
- Tested Applications: ELISA, Flow Cyt, WB
- Form: Liquid
- Storage Buffer: pH 8, containing 0.1% Sodium azide, 50% Glycerol, 0.61% Tris, and 0.01% Magnesium chloride
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Protein-based Substrate |
| Target Molecule | Avidin |
| Shipping Condition | Blue Ice |
| Label or Dye | Alkaline Phosphatase |
| Buffer | 50% glycerol, 0.61% Tris, 0.01% magnesium chloride, 0.1% sodium azide, pH8 |
| Content And Storage | Store at -20°C. |
| Form | Liquid |
| Detection Method | Colorimetric |
| Formulation | 50% glycerol, 0.61% Tris, 0.01% magnesium chloride, 0.1% sodium azide, pH 8 |
| Quantity | 1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.