Learn More
Abcam Native Chicken Deglycosylated Avidin Protein, Cy3, >95% Purity, 100 μg
Description
- Source: Chicken
- Expression System: Native
- Purity: >95%
- Applications: ELISA, Flow Cyt, WB
- Conjugation: Cy3™
- Formulation: Liquid. PBS, pH 7.3, 0.1% Sodium azide
- Storage Conditions: Store at +4°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Protein-based Substrate |
| Target Molecule | Avidin |
| Shipping Condition | Blue Ice |
| Buffer | PBS, 0.1% sodium azide, pH7.3 |
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C. |
| Form | Liquid |
| Label or Dye | Cy3 |
| Detection Method | Fluorescence |
| Formulation | PBS, 0.1% Sodium azide, pH 7.3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.