Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Native Chlamydia Pneumonia COMC Protein, Full Length, 100 μg

Catalog No. 502682849
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-268-2849 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-268-2849 Supplier Abcam Supplier No. AB274663100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Native full-length protein suitable for ELISA applications.

  • Protein Type: Native Full Length protein
  • Applications: ELISA
  • Species Reactivity: Bacteria (Chlamydia pneumonia)
  • Formulation: Liquid, pH 9.5
  • Storage Buffer Constituents: 0.75% Glycine, 0.15% (R*,R*)-1,4-Dimercaptobutan-2,3-diol

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Chlamydia Pneumonia COMC
Reactivity Bacteria
Shipping Condition Dry Ice
Form Liquid
Label or Dye Unconjugated
Buffer 0.75% Glycine, 0.15% (R*,R*)-1,4-dimercaptobutan-2,3-diol, pH9.5
Content And Storage Store at -80°C.
Cross Reactivity Chlamydia pneumonia
Formulation 0.75% Glycine, 0.15% (R*,R*)-1,4-Dimercaptobutan-2,3-diol, pH 9.5
Quantity 100 μg
For Use With (Application) ELISA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.