Learn More
Abcam Native Cow Acid Soluble Collagen Type 1
Supplier:  Abcam AB2819741ML
Encompass_Preferred
Native Cow Acid Soluble Collagen Type 1 is a Full Length protein, expressed in Native, with >95% purity, < 0.5 EU/mL endotoxin level and suitable for SDS-PAGE, Spectrum.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-1746
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.