Learn More
Abcam Native Cow Casein protein, 1MG
Supplier:  Abcam AB910921MG
Encompass_Preferred
Native Cow Casein protein is a Cow Full Length protein, expressed in Native, with >95% purity and suitable for SDS-PAGE, WB.
Protein/ Peptide/ Enzyme Synonyms: Alpha-S1-casein, CSN1S1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-9057