Learn More
Abcam Native human C1q protein (Active), 1MG
Supplier:  Abcam AB2828581MG
Encompass_Preferred
Native human C1q protein (Active) is a Human Full Length Complement Component 1q (C1q) protein with >98% purity and suitable for SDS-PAGE and Functional studies. The predicted molecular weight of ab282858 native protein is 410 kDa. - Save time and ensure accurate results - use the native C1q protein as a control - Optimal bioactivity and stability
Protein/ Peptide/ Enzyme Synonyms: Complement C1q subcomponent subunit A, C1QA
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2182512
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.