Learn More
Abcam Native Human Collagen VI protein, 500UG
Supplier:  Abcam AB7538500UG
Encompass_Preferred
Native Human Collagen VI protein is a Human Full Length protein with >95% and suitable for ELISA, IP, IHC, SDS-PAGE and western blot. - Save time and ensure accurate results - use our Collagen VI (COL6A1-COL6A3) protein as a control -Does not cross react with other collagens
Protein/ Peptide/ Enzyme Synonyms: Collagen alpha-1(VI) chain, COL6A1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-9825
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.