Learn More
Abcam Native Human Factor VII protein, 20UG
Supplier:  Abcam AB6238620UG
Encompass_Preferred
Native Human Factor VII protein is a Human Full Length protein, expressed in Native, with >95% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Coagulation factor VII, Proconvertin, Serum prothrombin conversion accelerator, SPCA, F7
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-271-0248