Learn More
Abcam Native Human Factor XIIIa protein (Active), 50UG
Supplier:  Abcam AB6241250UG
Encompass_Preferred
Native Human Factor XIIIa protein (Active) is a Human Full Length protein, expressed in Native, with >95% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: F13A, F13A1, Coagulation factor XIII A chain, Coagulation factor XIIIa, Protein-glutamine gamma-glutamyltransferase A chain, Transglutaminase A chain
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-271-0242